Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col4a5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163155.1 | NP_001156627.1 | 13 | 1058 | ||
NM_007736.4 | NP_031762.2 | 14 | 886 | |||
GenBank mRNA | AB041350.1 | - | 13 | 916 | ||
AK133179.1 | - | 14 | 887 | |||
BC082788.1 | - | 13 | 897 | 5343488 |