Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007738.3 | NP_031764.2 | 3 | 338 | ||
GenBank mRNA | U32107.1 | - | 3 | 338 |