Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col8a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007739.2 | NP_031765.2 | 4 | 2247 | ||
GenBank mRNA | AK018742.1 | - | 4 | 2226 | ||
AK029203.1 | - | 4 | 2221 | |||
AK032048.1 | - | 4 | 2226 | |||
AK046370.1 | - | 4 | 2226 | |||
AK159542.1 | - | 4 | 2222 | |||
AV334655.2 | - | 1 | 282 | |||
BC011061.1 | - | 4 | 2214 | 3674027 | ||
BC058281.1 | - | 4 | 2231 |