Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col9a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007740.3 | NP_031766.3 | 4 | 497 | ||
GenBank mRNA | AK031071.1 | - | 4 | 407 | ||
BC046970.1 | - | 4 | 385 | 6314986 | ||
CX207941.1 | - | 4 | 497 | |||
D17511.1 | - | 4 | 396 | |||
L12215.1 | - | 3 | 252 |