Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col9a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007741.2 | NP_031767.2 | 32 | 2236 | ||
GenBank mRNA | BC029697.1 | - | 32 | 2175 | 5355825 | |
BQ964231.1 | - | 3 | 474 |