Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007742.3 | NP_031768.2 | 40 | 2930 | ||
GenBank mRNA | AK132180.1 | - | 40 | 2951 | ||
AK159283.1 | - | 40 | 2948 | |||
AK159285.1 | - | 40 | 2952 | |||
BC003198.1 | - | 3 | 240 | |||
BC042642.1 | - | 33 | 2500 | |||
BC050014.1 | - | 40 | 2930 | 4225038 | ||
M14423.1 | - | 18 | 1280 | |||
U08020.1 | - | 40 | 2831 |