Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276248.1 | NP_001263177.1 | 2 | 474 | ||
NM_001276250.1 | NP_001263179.1 | 2 | 474 | |||
NM_007752.3 | NP_031778.2 | 2 | 474 | |||
GenBank mRNA | AK014835.1 | - | 2 | 460 | ||
AK043248.1 | - | 2 | 418 | |||
AK149523.1 | - | 2 | 310 | |||
BC062957.1 | - | 2 | 474 | 30547760 | ||
DQ336396.1 | - | 2 | 472 | |||
U49430.1 | - | 2 | 349 |