Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007754.2 | NP_031780.2 | 2 | 927 | ||
GenBank mRNA | AK032306.1 | - | 2 | 420 | ||
AK134907.1 | - | 3 | 780 | |||
AK137737.1 | - | 2 | 906 | |||
AK142583.1 | - | 2 | 897 | |||
D85391.1 | - | 2 | 915 |