Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007754.2 | NP_031780.2 | 2, 3 | 1041 | ||
GenBank mRNA | AK134907.1 | - | 3, 4 | 894 | ||
AK137737.1 | - | 2, 3 | 1020 | |||
AK142583.1 | - | 2, 3 | 1011 | |||
D85391.1 | - | 2, 3 | 1029 |