Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111112.1 | NP_001104582.1 | 2 | 413 | ||
NM_001312646.1 | NP_001299575.1 | 2 | 413 | |||
NM_001312647.1 | NP_001299576.1 | 2 | 325 | |||
NM_005339.4 | NP_005330.1 | 2 | 413 | |||
GenBank mRNA | AB022435.1 | - | 2 | 215 | ||
AK093533.1 | - | 2 | 234 | |||
AK295846.1 | - | 2 | 348 | |||
AK315524.1 | - | 2 | 339 | |||
BC022804.1 | - | 2 | 289 | 4893778 | ||
BC050600.1 | - | 2 | 267 | 6502901 | ||
CX786829.1 | - | 2 | 252 | |||
DA526283.1 | - | 2 | 413 | |||
U58522.1 | - | 2 | 132 |