Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301228.1 | NP_001288157.1 | 2 | 319 | ||
NM_001301229.1 | NP_001288158.1 | 2 | 210 | |||
NM_001301231.1 | NP_001288160.1 | 2 | 249 | |||
NM_005342.3 | NP_005333.2 | 2 | 214 | |||
GenBank mRNA | BC010378.1 | - | 1 | 134 | ||
BC037210.1 | - | 1 | 119 | |||
BC070482.1 | - | 3 | 298 | 6140626 | ||
BF979310.1 | - | 2 | 255 | |||
BX537505.1 | - | 2 | 199 | |||
DB191695.1 | - | 2 | 199 | |||
DN996967.1 | - | 2 | 221 | |||
L05085.1 | - | 1 | 119 | |||
Y10043.1 | - | 2 | 193 |