Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113529.1 | NP_001107001.1 | 2 | 497 | ||
NM_001113530.1 | NP_001107002.1 | 2 | 497 | |||
NM_007778.4 | NP_031804.3 | 2 | 497 | |||
GenBank mRNA | AK136808.1 | - | 2 | 271 | ||
AK138489.1 | - | 2 | 271 | |||
AK154261.1 | - | 2 | 498 | |||
AK154872.1 | - | 2 | 271 | |||
AK160995.1 | - | 2 | 271 | |||
AK166370.1 | - | 2 | 270 | |||
AK170154.1 | - | 2 | 207 | |||
BC025593.1 | - | 2 | 283 | |||
BC066187.1 | - | 2 | 251 | 30106218 | ||
BC066200.1 | - | 2 | 251 | 30100524 | ||
BC066205.1 | - | 2 | 251 | 30100601 | ||
CA328312.1 | - | 2 | 437 | |||
M15692.1 | - | 2 | 187 | |||
M21149.1 | - | 2 | 160 | |||
M21952.1 | - | 2 | 250 | |||
S78392.1 | - | 2 | 250 | |||
X05010.1 | - | 2 | 272 |