Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113529.1 | NP_001107001.1 | 4 | 701 | ||
NM_001113530.1 | NP_001107002.1 | 4 | 701 | |||
NM_007778.4 | NP_031804.3 | 4 | 701 | |||
GenBank mRNA | AK136808.1 | - | 4 | 475 | ||
AK138489.1 | - | 4 | 475 | |||
AK154261.1 | - | 4 | 702 | |||
AK154872.1 | - | 4 | 475 | |||
AK160995.1 | - | 4 | 475 | |||
AK166370.1 | - | 4 | 474 | |||
AK170154.1 | - | 4 | 411 | |||
BC025593.1 | - | 4 | 487 | |||
BC066187.1 | - | 4 | 455 | 30106218 | ||
BC066200.1 | - | 4 | 455 | 30100524 | ||
BC066205.1 | - | 4 | 455 | 30100601 | ||
M21149.1 | - | 4 | 364 | |||
M21952.1 | - | 4 | 454 | |||
S78392.1 | - | 4 | 454 | |||
X05010.1 | - | 4 | 476 |