Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf2rb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287389.1 | NP_001274318.1 | 12 | 4322 | ||
NM_007781.3 | NP_031807.1 | 14 | 4649 | |||
GenBank mRNA | M29855.1 | - | 14 | 4525 |