Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk2a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007788.3 | NP_031814.2 | 13 | 2111 | ||
GenBank mRNA | AK146032.1 | - | 13 | 2070 | ||
AK162515.1 | - | 1 | 186 | |||
BC026149.1 | - | 13 | 2016 | 4188101 | ||
BC060742.1 | - | 13 | 2064 | 4945773 | ||
BC089343.1 | - | 13 | 1988 | 30360017 |