Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007798.3 | NP_031824.1 | 4 | 385 | ||
GenBank mRNA | AK002456.1 | - | 4 | 334 | ||
AK083393.1 | - | 4 | 385 | |||
AK147192.1 | - | 4 | 332 | |||
AK149884.1 | - | 4 | 334 | |||
AK151790.1 | - | 4 | 332 | |||
AK153047.1 | - | 4 | 331 | |||
AK159992.1 | - | 4 | 370 | |||
AK167361.1 | - | 4 | 366 | |||
AK170959.1 | - | 4 | 358 | |||
AK190209.1 | - | 3 | 183 | |||
AK190539.1 | - | 3 | 183 | |||
AK191055.1 | - | 3 | 183 | |||
AK191391.1 | - | 3 | 183 | |||
AK196955.1 | - | 3 | 183 | |||
AK198213.1 | - | 3 | 183 | |||
AK199268.1 | - | 3 | 184 | |||
AK201893.1 | - | 3 | 183 | |||
AK202291.1 | - | 3 | 183 | |||
AK203639.1 | - | 3 | 183 | |||
BC006656.1 | - | 4 | 376 | 3500700 | ||
M14222.1 | - | 3 | 290 | |||
S69034.1 | - | 4 | 381 |