Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252572.1 | NP_001239501.1 | 11 | 1151 | ||
NM_007803.5 | NP_031829.2 | 12 | 1262 | |||
GenBank mRNA | AK084249.1 | - | 11 | 961 | ||
AK146856.1 | - | 12 | 1069 | |||
AK148010.1 | - | 12 | 1065 | |||
AK165555.1 | - | 11 | 954 | |||
AK168699.1 | - | 12 | 1063 | |||
AK168871.1 | - | 11 | 952 | |||
AK168945.1 | - | 11 | 962 | |||
BC011434.1 | - | 11 | 944 | 3981559 | ||
BC117820.1 | - | 11 | 964 | |||
U03184.1 | - | 12 | 1051 |