Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007806.3 | NP_031832.2 | 4 | 332 | ||
GenBank mRNA | AK021200.1 | - | 4 | 320 | ||
AK078854.1 | - | 4 | 303 | |||
AK131580.1 | - | 4 | 301 | |||
AK152414.1 | - | 4 | 323 | |||
AK171748.1 | - | 4 | 301 | |||
BC026791.1 | - | 4 | 283 | 3980538 | ||
BC060635.1 | - | 3 | 4338 | |||
BY005999.1 | - | 4 | 335 | |||
CF738398.1 | - | 4 | 315 | |||
EU791539.1 | - | 4 | 262 | |||
M31775.1 | - | 4 | 255 |