Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp26a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007811.2 | NP_031837.2 | 2, 3 | 515 | ||
GenBank mRNA | AF115769.1 | - | 2, 3 | 439 | ||
BC012673.1 | - | 2, 3 | 475 | 4210893 | ||
CB574526.1 | - | 1, 2 | 89 | |||
CK626921.1 | - | 2, 3 | 515 | |||
Y12657.1 | - | 2, 3 | 469 |