Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007824.2 | NP_031850.2 | 2 | 162 | ||
GenBank mRNA | AK050020.1 | - | 2 | 162 | ||
AK050210.1 | - | 2 | 163 | |||
AK050220.1 | - | 2 | 162 | |||
AK050260.1 | - | 2 | 162 | |||
BC130261.1 | - | 2 | 132 | 40142789 |