Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007842.2 | NP_031868.2 | 5 | 588 | ||
GenBank mRNA | AK013423.1 | - | 5 | 592 | ||
AK038298.1 | - | 1 | 1640 | |||
AK141824.1 | - | 5 | 583 | |||
AK148423.1 | - | 5 | 584 | |||
AK169735.1 | - | 5 | 588 | |||
BC033911.1 | - | 5 | 591 | |||
BC089159.1 | - | 5 | 582 | |||
U91922.1 | - | 5 | 545 |