Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dffb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007859.4 | NP_031885.3 | 6, 7 | 880 | ||
GenBank mRNA | AB009377.1 | - | 6, 7 | 788 | ||
AK077619.1 | - | 6, 7 | 926 | |||
AK089267.1 | - | 6, 7 | 858 | |||
BC053052.1 | - | 6, 7 | 810 | 6403143 |