Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dio1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007860.3 | NP_031886.3 | 2 | 435 | ||
GenBank mRNA | AK002549.1 | - | 2 | 436 | ||
BC019786.1 | - | 2 | 460 | 5133568 | ||
BC037080.1 | - | 2 | 471 | |||
BC089608.1 | - | 2 | 435 | 6766076 | ||
U49861.1 | - | 2 | 435 |