Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007868.5 | NP_031894.1 | 11 | 1513 | ||
GenBank mRNA | AK044536.1 | - | 11 | 1313 | ||
M18025.1 | - | 3 | 371 | |||
M68859.1 | - | 11 | 1486 |