Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007868.5 | NP_031894.1 | 37 | 5557 | ||
GenBank mRNA | M68859.1 | - | 37 | 5530 |