Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190817.1 | NP_001177746.1 | 11 | 1525 | ||
NM_007869.3 | NP_031895.1 | 11 | 1525 | |||
GenBank mRNA | AK133582.1 | - | 11 | 1507 | ||
AK144228.1 | - | 11 | 1482 | |||
BC080300.1 | - | 11 | 1506 | |||
L16953.1 | - | 11 | 1202 |