Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005388.4 | NP_005379.3 | 2 | 347 | ||
XM_017014785.1 | XP_016870274.1 | 2 | 305 | |||
GenBank mRNA | AF031463.1 | - | 2 | 324 | ||
AK313429.1 | - | 2 | 347 | |||
BC017227.2 | - | 2 | 222 | 3948368 |