Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXNB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163257.1 | NP_001156729.1 | 5 | 1560 | ||
NM_005393.2 | NP_005384.2 | 4 | 1394 | |||
GenBank mRNA | AB033032.1 | - | 4 | 1334 | ||
AF149019.1 | - | 4 | 1334 | |||
AK292535.1 | - | 4 | 1394 | |||
AK295762.1 | - | 4 | 1414 | |||
AK295849.1 | - | 2 | 213 | |||
AK295894.1 | - | 3 | 278 | |||
AK304705.1 | - | 4 | 459 | |||
AK307894.1 | - | 5 | 1537 |