Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290709.1 | NP_001277638.1 | 3, 4 | 437 | ||
NM_001290710.1 | NP_001277639.1 | 3, 4 | 437 | |||
NM_007897.3 | NP_031923.1 | 3, 4 | 437 | |||
GenBank mRNA | AK031239.1 | - | 3, 4 | 432 | ||
AK035781.1 | - | 3, 4 | 320 | |||
AK051438.1 | - | 3, 4 | 436 | |||
AK139021.1 | - | 3, 4 | 435 | |||
AK141175.1 | - | 3, 4 | 437 | |||
BC139362.1 | - | 3, 4 | 398 | 8862384 | ||
BC139364.1 | - | 3, 4 | 398 | 8862386 | ||
L12147.1 | - | 3, 4 | 403 |