Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009661.4 | NP_033791.1 | 11 | 1523 | ||
GenBank mRNA | AK028724.1 | - | 11 | 1519 | ||
AK158553.1 | - | 9 | 1397 | |||
BC015253.1 | - | 11 | 1523 | 4221815 | ||
U93277.1 | - | 11 | 1489 | |||
Y14696.1 | - | 11 | 1502 |