Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alox5ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK004002.1 | - | 2 | 181 | ||
AK075967.1 | - | 2 | 164 | |||
BC026209.1 | - | 2 | 168 | 1349819 | ||
BG085800.2 | - | 2 | 222 | |||
M96554.1 | - | 2 | 113 |