Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201413.1 | NP_001188342.1 | 17 | 2996 | ||
NM_001201414.1 | NP_001188343.1 | 16 | 2999 | |||
NM_001201415.1 | NP_001188344.1 | 16 | 2933 | |||
NM_001201416.1 | NP_001188345.1 | 7 | 1099 | |||
NM_009686.2 | NP_033816.1 | 18 | 3002 | |||
GenBank mRNA | AF133665.1 | - | 3 | 575 | ||
AK004792.1 | - | 18 | 2856 | |||
AK153260.1 | - | 16 | 2965 | |||
AK167649.1 | - | 17 | 2861 | |||
AK169959.1 | - | 16 | 2871 | |||
AK172405.1 | - | 7 | 1103 | |||
BC042797.1 | - | 8 | 1141 | |||
BC076587.1 | - | 16 | 2779 | 30550087 | ||
U70210.1 | - | 1 | 334 |