Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201413.1 | NP_001188342.1 | 17 | 3020 | ||
NM_001201414.1 | NP_001188343.1 | 16 | 3023 | |||
NM_001201415.1 | NP_001188344.1 | 16 | 2957 | |||
NM_001201416.1 | NP_001188345.1 | 7 | 1123 | |||
NM_009686.2 | NP_033816.1 | 18 | 3026 | |||
GenBank mRNA | AF133665.1 | - | 3 | 599 | ||
AK004792.1 | - | 18 | 2880 | |||
AK153260.1 | - | 16 | 2989 | |||
AK167649.1 | - | 17 | 2885 | |||
AK169959.1 | - | 16 | 2895 | |||
AK172405.1 | - | 7 | 1127 | |||
AY650904.1 | - | 1 | 42 | |||
BC042797.1 | - | 8 | 1165 | |||
BC076587.1 | - | 16 | 2803 | 30550087 | ||
U70210.1 | - | 1 | 358 |