Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277944.1 | NP_001264873.1 | 5 | 509 | ||
GenBank mRNA | AA059961.1 | - | 2 | 155 | ||
AK146468.1 | - | 4 | 310 | |||
BC024697.1 | - | 5 | 509 | 4195301 | ||
BG085960.2 | - | 4 | 352 | |||
CB949564.1 | - | 4 | 415 | |||
Z15090.1 | - | 4 | 310 |