Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009706.2 | NP_033836.2 | 1 | 867 | ||
GenBank mRNA | AK041016.1 | - | 2 | 899 | ||
BC150823.1 | - | 1 | 837 | 9087734 |