Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287530.1 | NP_001274459.1 | 4 | 683 | ||
NM_009707.4 | NP_033837.2 | 4 | 2051 | |||
NM_178754.3 | NP_848869.1 | 4 | 594 | |||
GenBank mRNA | AF012273.2 | - | 1173 | |||
AF177664.1 | - | 1173 | ||||
AK047827.1 | - | 4 | 595 | |||
AK048162.1 | - | 4 | 548 | |||
AK048507.1 | - | 4 | 549 | |||
AK077996.1 | - | 4 | 556 | |||
AK079651.1 | - | 4 | 507 | |||
AK164501.1 | - | 4 | 2052 | |||
AK166015.1 | - | 4 | 514 | |||
AK170575.1 | - | 4 | 550 | |||
AK171337.1 | - | 4 | 480 | |||
AK171677.1 | - | 4 | 2052 |