Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asgr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291131.1 | NP_001278060.1 | 9 | 1026 | ||
NM_001291132.1 | NP_001278061.1 | 8 | 1049 | |||
NM_009714.3 | NP_033844.1 | 9 | 1136 | |||
GenBank mRNA | AK132959.1 | - | 9 | 1027 | ||
BC014779.1 | - | 9 | 1042 | |||
BC022106.1 | - | 9 | 1098 | 5102469 | ||
U08372.1 | - | 8 | 1186 |