Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK002960.1 | - | 5 | 509 | ||
AK011312.1 | - | 5 | 523 | |||
AK019459.1 | - | 5 | 527 | |||
AK149176.1 | - | 5 | 495 | |||
AK150730.1 | - | 6 | 707 | |||
AK151250.1 | - | 6 | 707 | |||
AK151574.1 | - | 5 | 500 | |||
AK157544.1 | - | 5 | 563 | |||
AK158569.1 | - | 5 | 550 | |||
AK162311.1 | - | 5 | 492 | |||
AK167480.1 | - | 5 | 529 | |||
AK167626.1 | - | 5 | 500 | |||
AK168028.1 | - | 5 | 525 | |||
AK168041.1 | - | 5 | 526 | |||
AK168590.1 | - | 5 | 500 | |||
AK169137.1 | - | 5 | 527 | |||
AK169634.1 | - | 5 | 482 | |||
BC049640.1 | - | 5 | 457 | |||
BC087882.1 | - | 7 | 1011 | 6706159 | ||
BC089574.1 | - | 5 | 489 | 6756180 |