Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005439.2 | NP_005430.1 | 3 | 168 | ||
NR_026581.1 | 3 | 665 | ||||
GenBank mRNA | AF070539.1 | - | 3 | 298 | ||
AK289869.1 | - | 3 | 168 | |||
AK313998.1 | - | 3 | 469 | |||
BC000898.2 | - | 3 | 238 | 2900086 | ||
BC002340.2 | - | 3 | 278 | 2820309 | ||
BT019393.1 | - | 2 | 105 | |||
BT019432.1 | - | 2 | 105 | |||
CR541700.1 | - | 2 | 105 | |||
U57342.1 | - | 3 | 284 |