Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009734.3 | NP_033864.3 | 3 | 406 | ||
GenBank mRNA | AK133245.1 | - | 3 | 407 | ||
BC141108.1 | - | 3 | 346 | 9055720 | ||
BY724850.1 | - | 3 | 407 | |||
D43921.1 | - | 3 | 398 |