Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep131 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009734.3 | NP_033864.3 | 23 | 2910 | ||
GenBank mRNA | AK133245.1 | - | 23 | 2913 | ||
AK134381.1 | - | 15 | 2552 | |||
BC141108.1 | - | 23 | 2850 | 9055720 | ||
D43921.1 | - | 23 | 2905 |