Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171739.1 | NP_001165210.1 | 5 | 968 | ||
NM_009736.3 | NP_033866.4 | 5 | 968 | |||
GenBank mRNA | AF022223.1 | - | 5 | 909 | ||
AK009149.1 | - | 5 | 879 | |||
AK013611.1 | - | 3 | 254 | |||
AK154469.1 | - | 5 | 918 | |||
AK171094.1 | - | 5 | 918 | |||
BC003722.1 | - | 5 | 930 | 3495306 | ||
BC069918.1 | - | 5 | 905 | 4979705 | ||
BC093509.1 | - | 5 | 879 | 4978330 |