Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bag1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171739.1 | NP_001165210.1 | 6, 7 | 1078 | ||
NM_009736.3 | NP_033866.4 | 6, 7 | 1078 | |||
GenBank mRNA | AF022223.1 | - | 6, 7 | 1019 | ||
AK009149.1 | - | 6, 7 | 989 | |||
AK013611.1 | - | 4, 5 | 364 | |||
AK154469.1 | - | 6, 7 | 1027 | |||
AK171094.1 | - | 6, 7 | 1027 | |||
AK188292.1 | - | 2, 3 | 154 | |||
BC003722.1 | - | 6, 7 | 1040 | 3495306 | ||
BC069918.1 | - | 6, 7 | 1015 | 4979705 | ||
BC093509.1 | - | 6, 7 | 989 | 4978330 |