Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bckdk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009739.3 | NP_033869.1 | 2 | 425 | ||
GenBank mRNA | AF043070.1 | - | 1 | 266 | ||
AK075602.1 | - | 2 | 351 | |||
AK145479.1 | - | 2 | 396 | |||
AK150607.1 | - | 2 | 417 | |||
AK150723.1 | - | 2 | 417 | |||
AK167519.1 | - | 2 | 417 | |||
AK218245.1 | - | 1 | 134 | |||
BC046595.1 | - | 2 | 395 | 6313106 |