Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005208.4 | NP_005199.2 | 5 | 381 | ||
XM_017024198.1 | XP_016879687.1 | 6 | 677 | |||
GenBank mRNA | BC069537.1 | - | 5 | 374 | 7262224 | |
BC113693.1 | - | 5 | 375 | 8322745 | ||
KM230602.1 | - | 5 | 363 | |||
KM230603.1 | - | 5 | 362 | |||
KM230604.1 | - | 5 | 359 | |||
KM230605.1 | - | 4 | 359 | |||
KR336660.1 | - | 5 | 359 | |||
KR336661.1 | - | 2 | 48 | |||
U59058.1 | - | 3 | 247 |