Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110233.1 | NP_001103703.1 | 3 | 774 | ||
NM_001110234.1 | NP_001103704.1 | 3 | 733 | |||
NM_009750.2 | NP_033880.1 | 3 | 779 | |||
GenBank mRNA | AF097440.1 | - | 3 | 700 | ||
AK003294.1 | - | 3 | 687 | |||
AK004531.1 | - | 3 | 709 | |||
AK010500.1 | - | 3 | 706 | |||
AK214623.1 | - | 1 | 116 | |||
BC027815.1 | - | 3 | 705 | 5250429 | ||
CB055608.1 | - | 3 | 632 | |||
CF618290.1 | - | 3 | 723 |
Gene Symbol: LOC102642808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK219603.1 | - | 1 | 75 |