Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ngfrap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110233.1 | NP_001103703.1 | 3 | 595 | ||
NM_001110234.1 | NP_001103704.1 | 3 | 554 | |||
NM_009750.2 | NP_033880.1 | 3 | 600 | |||
GenBank mRNA | AF097440.1 | - | 3 | 521 | ||
AF187066.1 | - | 3 | 526 | |||
AK003294.1 | - | 3 | 508 | |||
AK004531.1 | - | 3 | 530 | |||
AK010500.1 | - | 3 | 527 | |||
BC027815.1 | - | 3 | 526 | 5250429 | ||
CB055608.1 | - | 3 | 453 | |||
CF618290.1 | - | 3 | 544 |