Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 2 | 208 | ||
GenBank mRNA | AK034049.1 | - | 2 | 187 | ||
AK042069.1 | - | 2 | 178 | |||
AK048460.1 | - | 2 | 139 | |||
AK076517.1 | - | 2 | 179 | |||
AK153218.1 | - | 2 | 191 | |||
AK166473.1 | - | 2 | 193 | |||
AK171593.1 | - | 2 | 211 | |||
BC028875.1 | - | 2 | 128 | 4240285 | ||
M57734.1 | - | 2 | 167 |