Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009752.2 | NP_033882.1 | 7, 8 | 912 | ||
GenBank mRNA | AK034049.1 | - | 7, 8 | 891 | ||
AK034966.1 | - | 713 | ||||
AK042069.1 | - | 7, 8 | 882 | |||
AK048460.1 | - | 7, 8 | 843 | |||
AK076517.1 | - | 7, 8 | 883 | |||
AK153218.1 | - | 7, 8 | 894 | |||
AK167271.1 | - | 5, 6 | 522 | |||
AK171593.1 | - | 7, 8 | 915 | |||
BC028875.1 | - | 7, 8 | 832 | 4240285 | ||
M57734.1 | - | 7, 8 | 871 |