Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009755.3 | NP_033885.2 | 18 | 2664 | ||
GenBank mRNA | AK004995.2 | - | 18 | 2298 | ||
AK147534.1 | - | 16 | 4275 | |||
BC003925.1 | - | 8 | 1688 | |||
BC053923.1 | - | 18 | 3250 | |||
BC063079.1 | - | 14 | 1794 | |||
BC066062.1 | - | 18 | 2592 | 6849066 | ||
L24755.1 | - | 18 | 2441 |